1-(Dicyanomethylene)-3-iminoisoindoline ≥98.0% (by total nitrogen basis)

Supplier: TCI America

D2080-1G
TCD2080-001GEA 209 USD
TCD2080-001G
1-(Dicyanomethylene)-3-iminoisoindoline ≥98.0% (by total nitrogen basis)
1-(Dicyanomethylene)-3-iminoisoindoline

CAS Number: 43002-19-3
MDL Number: MFCD00142610
Molecular Formula: C11H6N4
Molecular Weight: 194.20
Purity/Analysis Method: 98.0% (N)
Form: Crystal

Formula: C₁₁H₆N₄
MDL Number: MFCD00142610
CAS Number: 43002-19-3
https://us.vwr.com/stibo/web/std.lang.all/88/63/28978863.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (N)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR