2,5-Dimethyl-2-hexene ≥96.0%
Supplier: TCI America
D2076-10ML
TCD2076-010MLEA
270.88
USD
TCD2076-010ML
2,5-Dimethyl-2-hexene ≥96.0%
2,5-Dimethyl-2-hexene
CAS Number: 3404-78-2
MDL Number: MFCD00026436
Molecular Formula: C8H16
Molecular Weight: 112.22
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 113
Flash Point (°C): 11
Specific Gravity (20/20): 0.72
Formula:
C₈H₁₆ MW: 112.22 g/mol Boiling Pt: 113 °C Density: 0,72 (20 °C) Flash Pt: 11 °C |
MDL Number:
MFCD00026436 CAS Number: 3404-78-2 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...