TOKYD2065

D2065-25ML
TCD2065-025MLEA 90.49 USD
TCD2065-025ML
TOKYD2065
CAS Number: 57597-62-3
MDL Number: MFCD00044797
Molecular Formula: C5H9NO2
Molecular Weight: 115.13
Purity/Analysis Method: 93.0% (GC)
Form: Clear Liquid
Boiling point (°C): 195
Flash Point (°C): 83
Specific Gravity (20/20): 1.02

Other Product Names / Synonyms: Cyanoacetaldehyde Dimethyl Acetal
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR