Allyl-n-octyl ether ≥95.0%

Supplier: TCI America

A1052-10ML
TCA1052-10MLEA 130.88 USD
TCA1052-10ML
Allyl-n-octyl ether ≥95.0%
Allyl-n-octyl ether

CAS Number: 3295-97-4
MDL Number: MFCD00015294
Molecular Formula: C11H22O
Molecular Weight: 170.30
Purity/Analysis Method: 95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 206
Specific Gravity (20/20): 0.81

Formula: C₁₁H₂₂O
Boiling Pt: 206 °C
Density: 0.81 (20 °C)
MDL Number: MFCD00015294
CAS Number: 3295-97-4
https://us.vwr.com/stibo/web/std.lang.all/87/99/28928799.jpg

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR