TCIAD2055

D2055-1G
TCD2055-001GEA 121.34 USD
TCD2055-001G
TCIAD2055
MDL Number: MFCD00143993
Molecular Formula: C11H12N2
Molecular Weight: 172.23
Form: Crystal
Color: Slightly Pale Yellow
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR