1,2-Diacetoxybenzene ≥98.0%
Supplier: TCI America
Synonyms:
Catechol Diacetate
D2044-25G
TCD2044-025GEA
174.5
USD
TCD2044-025G
1,2-Diacetoxybenzene ≥98.0%
1,2-Diacetoxybenzene
CAS Number: 635-67-6
MDL Number: MFCD00017219
Molecular Formula: C10H10O4
Molecular Weight: 194.19
Purity/Analysis Method:
Form: Crystal
Boiling point (°C): 140
Melting point (°C): 64
Other Product Names / Synonyms: Catechol Diacetate
Formula:
C₁₀H₁₀O₄ MW: 194.19 g/mol Boiling Pt: 140 °C Melting Pt: 64 °C |
MDL Number:
MFCD00017219 CAS Number: 635-67-6 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...