Azobenzene-4-sulfonyl chloride ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 4-Phenylazobenzenesulfonyl Chloride4-Phenylazobenzenesulphonyl Chloride

A1050-5G
TCA1050-005GEA 240.48 USD
TCA1050-005G
Azobenzene-4-sulfonyl chloride ≥98.0% (by titrimetric analysis)
Azobenzene-4-sulfonyl chloride

CAS Number: 58359-53-8
MDL Number: MFCD00024883
Molecular Formula: C12H9ClN2O2S
Molecular Weight: 280.73
Purity/Analysis Method: 98.0% (T)
Form: Crystal
Melting point (°C): 122
Storage Temperature: 0-10°C


Other Product Names / Synonyms: 4-Phenylazobenzenesulfonyl Chloride

Formula: C₁₂H₉ClN₂O₂S
MW: 280.73 g/mol
Melting Pt: 122 °C
MDL Number: MFCD00024883
CAS Number: 58359-53-8
https://us.vwr.com/stibo/web/std.lang.all/87/97/28928797.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR