2'-(N,N-Dimethylamino)-5'-nitroacetanilide ≥98.0% (by titrimetric analysis)
Supplier: TCI America
D2020-1G
TCD2020-001GEA
174
USD
TCD2020-001G
2'-(N,N-Dimethylamino)-5'-nitroacetanilide ≥98.0% (by titrimetric analysis)
2'-(N,N-Dimethylamino)-5'-nitroacetanilide
MDL Number: MFCD00130122
Molecular Formula: C10H13N3O3
Molecular Weight: 223.23
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 166
Formula:
C₁₀H₁₃N₃O₃ Melting Pt: 166 °C |
MDL Number:
MFCD00130122 CAS Number: 5367-36-2 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...