1,3-Diiminobenz[f]isoindoline ≥95.0% (by total nitrogen basis)

Supplier: TCI America

D2009-1G
TCD2009-001GEA 231.25 USD
TCD2009-001G
1,3-Diiminobenz[f]isoindoline ≥95.0% (by total nitrogen basis)
1,3-Diiminobenz[f]isoindoline

CAS Number: 65558-69-2
MDL Number: MFCD00059067
Molecular Formula: C12H9N3
Molecular Weight: 195.23
Purity/Analysis Method: 95.0% (N)
Form: Crystal

Formula: C₁₂H₉N₃
MDL Number: MFCD00059067
CAS Number: 65558-69-2
https://us.vwr.com/stibo/web/std.lang.all/87/99/28978799.jpg

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (N)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR