Decahydro-2-naphthol (mixture of isomers) ≥95.0%
Supplier: TCI America
Synonyms:
Decalin-2-ol
D2000-25G
TCD2000-025GEA
61.92
USD
TCD2000-025G
Decahydro-2-naphthol (mixture of isomers) ≥95.0%
Decahydro-2-naphthol (mixture of isomers)
CAS Number: 825-51-4
MDL Number: MFCD00004132
Molecular Formula: C10H18O
Molecular Weight: 154.25
Purity/Analysis Method:
Form: Paste
Flash Point (°C): 112
Other Product Names / Synonyms: Decalin-2-ol
Formula:
C₁₀H₁₈O MW: 154.25 g/mol Flash Pt: 112 °C |
MDL Number:
MFCD00004132 CAS Number: 825-51-4 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...