Naphthalene-2,3-dicarbonitrile ≥98.0%

Supplier: TCI America

Synonyms: 2,3-Naphthalenedicarbonitrile2,3-Dicyanonaphthalene

D1977-10G D1977-1G
TCD1977-001GEA 84.21 USD
TCD1977-001G TCD1977-010G
Naphthalene-2,3-dicarbonitrile ≥98.0%
Naphthalene-2,3-dicarbonitrile

CAS Number: 22856-30-0
MDL Number: MFCD00059063
Molecular Formula: C12H6N2
Molecular Weight: 178.19
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Melting point (°C): 257


Product Categories: Materials Chemistry, Building Blocks for Phthalocyanines
Other Product Names / Synonyms: 2,3-Naphthalenedicarbonitrile

Formula: C₁₂H₆N₂
MW: 178.1894 g/mol
Melting Pt: 257 °C
MDL Number: MFCD00059063
CAS Number: 22856-30-0
https://us.vwr.com/stibo/web/std.lang.all/87/73/28978773.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR