3,4-Dimethyl-3-hexanol ≥99.0%

Supplier: TCI America

D1970-10ML
TCD1970-010MLEA 149.14 USD
TCD1970-010ML
3,4-Dimethyl-3-hexanol ≥99.0%
3,4-Dimethyl-3-hexanol

CAS Number: 19550-08-4
MDL Number: MFCD00021820
Molecular Formula: C8H18O
Molecular Weight: 130.23
Purity/Analysis Method: 99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 58
Specific Gravity (20/20): 0.85

Formula: C₈H₁₈O
Boiling Pt: 58 °C
Density: 0,85 (20 °C)
MDL Number: MFCD00021820
CAS Number: 19550-08-4

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR