1,3-Diisopropenylbenzene ≥97.0% stabilized

Supplier: TCI America

D1957-25ML
TCD1957-025MLEA 39.72 USD
TCD1957-025ML
1,3-Diisopropenylbenzene ≥97.0% stabilized
1,3-Diisopropenylbenzene

(stabilized with TBC)
CAS Number: 3748-13-8
MDL Number: MFCD00008584
Molecular Formula: C12H14
Molecular Weight: 158.24
Purity/Analysis Method: 97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 231
Melting point (°C): -40
Flash Point (°C): 93
Specific Gravity (20/20): 0.92
Storage Temperature: 0-10°C

Formula: C₁₂H₁₄
MW: 158.2396 g/mol
Boiling Pt: 231 °C
Melting Pt: –40 °C
Density: 0.92 (20 °C)
Flash Pt: 93 °C
MDL Number: MFCD00008584
CAS Number: 3748-13-8
https://us.vwr.com/stibo/web/std.lang.all/87/55/28978755.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR