Allyl dimethyldithiocarbamate ≥98.0%
Supplier: TCI America
Synonyms:
Dimethyldithiocarbamic Acid Allyl Ester
D1944-5G
TCD1944-005GEA
212
USD
TCD1944-005G
Allyl dimethyldithiocarbamate ≥98.0%
Allyl dimethyldithiocarbamate
CAS Number: 20821-66-3
MDL Number: MFCD00059057
Molecular Formula: C6H11NS2
Molecular Weight: 161.28
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 108
Specific Gravity (20/20): 1.11
Product Categories: Synthetic Organic Chemistry, Thiocarbonyl Compounds
Other Product Names / Synonyms: Dimethyldithiocarbamic Acid Allyl Ester
Formula:
C₆H₁₁NS₂ MW: 161.28 g/mol Boiling Pt: 108 °C Density: 1.11 (20 °C) |
MDL Number:
MFCD00059057 CAS Number: 20821-66-3 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...