Dodecanal dimethyl acetal ≥97.0%
Supplier: TCI America
Synonyms:
1,1-Dimethoxydodecane
, Lauryl Aldehyde Dimethyl Acetal
D1937-25ML
TCD1937-025MLEA
104.5
USD
TCD1937-025ML
Dodecanal dimethyl acetal ≥97.0%
Dodecanal dimethyl acetal
CAS Number: 14620-52-1
MDL Number: MFCD00036645
Molecular Formula: C14H30O2
Molecular Weight: 230.39
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 133
Specific Gravity (20/20): 0.85
Other Product Names / Synonyms: Lauryl Aldehyde Dimethyl Acetal, 1,1-Dimethoxydodecane
Formula:
C₁₄H₃₀O₂ MW: 230.39 g/mol Boiling Pt: 133 °C Density: 0.85 (20 °C) |
MDL Number:
MFCD00036645 CAS Number: 14620-52-1 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...