Diethylcarbamazine citrate ≥98.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
N,N-Diethyl-4-methyl-1-piperazinecarboxamide Citrate
D1898-10G
TCD1898-010GEA
242.51
USD
TCD1898-010G
Diethylcarbamazine citrate ≥98.0% (by titrimetric analysis)
Diethylcarbamazine citrate
CAS Number: 1642-54-2
MDL Number: MFCD00039124
Molecular Formula: C16H29N3O8
Molecular Weight: 391.42
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 140
Product Categories: Bioscience, Anthelmintic Ingredients
Other Product Names / Synonyms: N,N-Diethyl-4-methyl-1-piperazinecarboxamide Citrate
Formula:
C₁₆H₂₉N₃O₈ MW: 391.42 g/mol Melting Pt: 140 °C |
MDL Number:
MFCD00039124 CAS Number: 1642-54-2 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...