Ethylene glycolbis(3-aminopropyl)ether ≥98.0% (by GC, titration analysis)
Supplier: TCI America
Synonyms:
1,2-Bis(3-aminopropoxy)ethane
D1840-5ML
D1840-500ML
D1840-25ML
TCD1840-5MLEA
16.24
USD
TCD1840-5ML
TCD1840-500ML
TCD1840-025ML
Ethylene glycolbis(3-aminopropyl)ether ≥98.0% (by GC, titration analysis)
Ethylene glycolbis(3-aminopropyl)ether
CAS Number: 2997-01-5
MDL Number: MFCD00059938
Molecular Formula: C8H20N2O2
Molecular Weight: 176.26
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 96
Specific Gravity (20/20): 0.98
Other Product Names / Synonyms: 1,2-Bis(3-aminopropoxy)ethane
Formula:
C₈H₂₀N₂O₂ MW: 176.26 g/mol Boiling Pt: 96 °C Density: 0.98 (20 °C) |
MDL Number:
MFCD00059938 CAS Number: 2997-01-5 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...