1,3-Divinyl-1,1,3,3-tetramethyldisilazane ≥95.0%
Supplier: TCI America
Synonyms:
1,1,3,3-Tetramethyl-1,3-divinyldisilazane
D1769-5ML
TCD1769-005MLEA
21.32
USD
TCD1769-005ML
1,3-Divinyl-1,1,3,3-tetramethyldisilazane ≥95.0%
1,3-Divinyl-1,1,3,3-tetramethyldisilazane
CAS Number: 7691-02-3
MDL Number: MFCD00042846
Molecular Formula: C8H19NSi2
Molecular Weight: 185.42
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 160
Flash Point (°C): 42
Specific Gravity (20/20): 0.82
Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: 1,1,3,3-Tetramethyl-1,3-divinyldisilazane
Formula:
C₈H₁₉NSi₂ MW: 185.42 g/mol Boiling Pt: 160 °C Density: 0.82 (20 °C) Flash Pt: 42 °C |
MDL Number:
MFCD00042846 CAS Number: 7691-02-3 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...