N-(2,6-Diethylphenylcarbamoylmethyl)iminodiacetic acid ≥99.0% (by HPLC, titration analysis)
Supplier: TCI America
D1735-5G
TCD1735-005GEA
162
USD
TCD1735-005G
N-(2,6-Diethylphenylcarbamoylmethyl)iminodiacetic acid ≥99.0% (by HPLC, titration analysis)
N-(2,6-Diethylphenylcarbamoylmethyl)iminodiacetic acid
CAS Number: 63245-28-3
MDL Number: MFCD00059631
Molecular Formula: C16H22N2O5
Molecular Weight: 322.36
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 190
Formula:
C₁₆H₂₂N₂O₅ Melting Pt: 190 °C |
MDL Number:
MFCD00059631 CAS Number: 63245-28-3 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...