9,10-Dicyanoanthracene ≥98.0% (by HPLC, total nitrogen)
Supplier: TCI America
Synonyms:
9,10-Anthracenedicarbonitrile
, Anthracene-9,10-dicarbonitrile
D1656-1G
D1656-5G
TCD1656-005GEA
269
USD
TCD1656-005G
TCD1656-001G
9,10-Dicyanoanthracene ≥98.0% (by HPLC, total nitrogen)
9,10-Dicyanoanthracene
CAS Number: 1217-45-4
MDL Number: MFCD00041733
Molecular Formula: C16H8N2
Molecular Weight: 228.25
Purity/Analysis Method:
Form: Crystal
Other Product Names / Synonyms: Anthracene-9,10-dicarbonitrile
Formula:
C₁₆H₈N₂ MW: 228.25 g/mol |
MDL Number:
MFCD00041733 CAS Number: 1217-45-4 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...