Dipropyl sulfate ≥90.0%

Supplier: TCI America
Danger

D1638-500G D1638-25G
TCD1638-500GEA 298.32 USD
TCD1638-500G TCD1638-025G
Dipropyl sulfate ≥90.0%
Dipropyl sulphate

[Alkylating Agent]
CAS Number: 598-05-0
MDL Number: MFCD00059243
Molecular Formula: C6H14O4S
Molecular Weight: 182.23
Purity/Analysis Method: 90.0% (GC)
Form: Clear Liquid
Boiling point (°C): 115
Specific Gravity (20/20): 1.11

Formula: C₆H₁₄O₄S
MW: 182.24 g/mol
Boiling Pt: 115 °C
Density: 1.11 (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00059243
CAS Number: 598-05-0
UN: 3265
ADR: 8,II
https://us.vwr.com/stibo/web/std.lang.all/84/34/28978434.jpg

Order Now

Specification Test Results

Purity/Analysis Method 90.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR