Disperse diazo black 3BF
D1615-25G
TCD1615-025GEA
178.19
USD
TCD1615-025G
Disperse diazo black 3BF
Disperse diazo black 3BF
CAS Number: 6232-57-1
MDL Number: MFCD00059857
Molecular Formula: C14H16N4O
Molecular Weight: 256.31
Form: Crystal
Color: Yellow
Formula:
C₁₄H₁₆N₄O |
MDL Number:
MFCD00059857 CAS Number: 6232-57-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...