3,6-Dihydroxyphthalonitrile ≥97.0% (by GC, titration analysis)
Supplier: TCI America
|
Warning
|
Synonyms:
2,3-Dicyanohydroquinone
D1610-25G
TCD1610-025GEA
98.46
USD
TCD1610-025G
3,6-Dihydroxyphthalonitrile ≥97.0% (by GC, titration analysis)
3,6-Dihydroxyphthalonitrile
CAS Number: 4733-50-0
MDL Number: MFCD00001790
Molecular Formula: C8H4N2O2
Molecular Weight: 160.13
Purity/Analysis Method:
Form: Crystal
Product Categories: Materials Chemistry, Building Blocks for Phthalocyanines
Other Product Names / Synonyms: 3,6-Dihydroxyphthalonitrile
Formula:
C₈H₄N₂O₂ MW: 160.13 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00001790 CAS Number: 4733-50-0 EINECS: 225-241-0 UN: 3439 ADR: 6.1,II |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...