2,6-Dimethylcyclohexanol (mixture of isomers) ≥99.0%

Supplier: TCI America

Synonyms: 2,6-Dimethylcyclohexanol

D1578-25ML
TCD1578-025MLEA 45 USD
TCD1578-025ML
2,6-Dimethylcyclohexanol (mixture of isomers) ≥99.0%
2,6-Dimethylcyclohexanol (mixture of isomers)

CAS Number: 5337-72-4
MDL Number: MFCD00001502
Molecular Formula: C8H16O
Molecular Weight: 128.22
Purity/Analysis Method: 99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 173
Flash Point (°C): 55
Specific Gravity (20/20): 0.92

Formula: C₈H₁₆O
Boiling Pt: 173 °C
Density: 0.92 (20 °C)
Flash Pt: 55 °C
MDL Number: MFCD00001502
CAS Number: 5337-72-4
https://us.vwr.com/stibo/web/std.lang.all/83/81/28978381.jpg

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR