(±)-γ-Dodecalactone ≥97.0%

Supplier: TCI America

Synonyms: (±)-Dihydro-5-octylfuran-2(3H)-onegamma-Dodecalactonegamma-Dodecanolactonegamma-Laurolactone(±)-Dodecan-4-olide5-Octyldihydrofuran-2(3H)-one

D1576-25ML
TCD1576-025MLEA 80.81 USD
TCD1576-025ML
(±)-γ-Dodecalactone ≥97.0%
(±)-γ-Dodecalactone

CAS Number: 2305-05-7
MDL Number: MFCD00036499
Molecular Formula: C12H22O2
Molecular Weight: 198.31
Purity/Analysis Method: 97.0% (GC)
Form: Clear Liquid
Flash Point (°C): 110
Specific Gravity (20/20): 0.94


Product Categories: Bioscience, Lipids
Other Product Names / Synonyms: gamma-Dodecalactone, gamma-Laurolactone

Formula: C₁₂H₂₂O₂
MW: 198.31 g/mol
Density: 0.94 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00036499
CAS Number: 2305-05-7
EINECS: 218-971-6
https://us.vwr.com/stibo/web/std.lang.all/83/79/28978379.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR