4,7,10-Trioxa-1,13-tridecanediamine ≥98.0% (by GC, titration analysis)
Supplier: TCI America
Synonyms:
3,3'-((Oxybis(ethane-2,1-diyl))bis(oxy))bis(propan-1-amine)
, 1,13-Diamino-4,7,10-trioxatridecane
CAS Number: 4246-51-9
MDL Number: MFCD00059850
Molecular Formula: C10H24N2O3
Molecular Weight: 220.31
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 151
Flash Point (°C): 166
Specific Gravity (20/20): 1.01
Product Categories: Materials Chemistry, Monomers, Bioscience, Crosslinkers, PEGylation Reagents & PEG Spacers
Other Product Names / Synonyms: Bis[2-(3-aminopropoxy)ethyl] Ether, 4,7,10-Trioxa-1,13-tridecanediamine
Formula:
C₁₀H₂₄N₂O₃ MW: 220.3092 g/mol Boiling Pt: 151 °C Density: 1.01 (20 °C) Flash Pt: 166 °C |
MDL Number:
MFCD00059850 CAS Number: 4246-51-9 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...