7-Acetoxy-4-bromomethylcoumarin ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

A0979-1G
TCA0979-001GEA 158.29 USD
TCA0979-001G
7-Acetoxy-4-bromomethylcoumarin ≥98.0% (by HPLC, titration analysis)
7-Acetoxy-4-bromomethylcoumarin

[for HPLC Labeling]
CAS Number: 2747-04-8
MDL Number: MFCD00010707
Molecular Formula: C12H9BrO4
Molecular Weight: 297.10
Purity/Analysis Method: 98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 182

Formula: C₁₂H₉BrO₄
Melting Pt: 182 °C
MDL Number: MFCD00010707
CAS Number: 2747-04-8
https://us.vwr.com/stibo/web/std.lang.all/87/28/28928728.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR