Acetyl sulfide ≥95.0%

Supplier: TCI America

Synonyms: Diacetyl SulphideDiacetyl sulfide

A0976-5G A0976-25G
TCA0976-025GEA 711.3 USD
TCA0976-025G TCA0976-005G
Acetyl sulfide ≥95.0%
Acetyl sulfide

CAS Number: 3232-39-1
MDL Number: MFCD00008753
Molecular Formula: C4H6O2S
Molecular Weight: 118.15
Purity/Analysis Method: 95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 120
Flash Point (°C): 27
Specific Gravity (20/20): 1.13


Other Product Names / Synonyms: Diacetyl Sulfide

Formula: C₄H₆O₂S
MW: 118.15 g/mol
Boiling Pt: 120 °C
Density: 1.13 (20 °C)
Flash Pt: 27 °C
MDL Number: MFCD00008753
CAS Number: 3232-39-1
https://us.vwr.com/stibo/web/std.lang.all/87/26/28928726.jpg

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR