Acriflavine ≥70.0% (by HPLC), crystals
Supplier: TCI America
|
Danger
|
Synonyms:
3,6-Diamino-10-methylacridinium chloride-3,6-acridinediamine mixt
A0134-25G
TCA0134-025GEA
51.77
USD
TCA0134-025G
Acriflavine ≥70.0% (by HPLC), crystals
Acriflavine
Mixture of 3,6-Diamino-10-methylacridinium Chloride and 3,6-Diaminoacridine
Formula:
C₂₇H₂₅ClN₆ MW: 468.9806 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00064307 CAS Number: 8048-52-0 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...