4,4'-Dipyridyl sulfide ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: Di(pyridin-4-yl)sulfane4,4'-ThiodipyridineDi(pyridin-4-yl)sulphane

D1483-5G
TCD1483-005GEA 402 USD
TCD1483-005G
4,4'-Dipyridyl sulfide ≥98.0% (by GC, titration analysis)
4,4'-Dipyridyl sulfide

CAS Number: 37968-97-1
MDL Number: MFCD00059784
Molecular Formula: C10H8N2S
Molecular Weight: 188.25
Purity/Analysis Method: 98.0% (GC,T)
Form: Crystal
Melting point (°C): 69


Other Product Names / Synonyms: 4,4'-Thiodipyridine

Formula: C₁₀H₈N₂S
MW: 188.25 g/mol
Melting Pt: 69 °C
MDL Number: MFCD00059784
CAS Number: 37968-97-1
https://us.vwr.com/stibo/web/std.lang.all/83/08/28978308.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR