Direct yellow 26

Supplier: TCI America

Synonyms: Direct Fast Yellow 5GLDirect Yellow MCAmanil Fast Yellow 5GL

D1473-25G
TCD1473-025GEA 85 USD
TCD1473-025G
Direct yellow 26
Direct yellow 26

CAS Number: 2829-42-7
MDL Number: MFCD00266524
Molecular Formula: C27H20N6O7
Molecular Weight: 584.46
Form: Crystal
Color: Pale Yellow


Other Product Names / Synonyms: Amanil Fast Yellow 5GL, Direct Fast Yellow 5GL, Direct Yellow MC

Formula: C₂₇H₂₀N₆O₇
MW: 584.46 g/mol
MDL Number: MFCD00266524
CAS Number: 2829-42-7
https://us.vwr.com/stibo/web/std.lang.all/83/00/28978300.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR