Direct fast brown BX
Synonyms:
Direct Brown B
, Calcomine Brown BX
D1471-25G
TCD1471-025GEA
66.6
USD
TCD1471-025G
Direct fast brown BX
Direct fast brown BX
CAS Number: 3476-90-2
MDL Number: MFCD00059538
Molecular Formula: C35H25N5O7S
Molecular Weight: 703.64
Form: Crystal
Color: Gray
Other Product Names / Synonyms: Calcomine Brown BX, Direct Brown B
Formula:
C₃₅H₂₅N₅O₇S MW: 703.64 g/mol |
MDL Number:
MFCD00059538 CAS Number: 3476-90-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...