Direct fast brown M
Synonyms:
Direct Brown M
D1470-25G
TCD1470-025GEA
30.9
USD
TCD1470-025G
Direct fast brown M
Direct fast brown M
CAS Number: 2429-82-5
MDL Number: MFCD00059539
Molecular Formula: C29H21N5O7S
Molecular Weight: 627.54
Form: Crystal
Color: Pale Yellow
Other Product Names / Synonyms: Direct Brown M
Formula:
C₂₉H₂₁N₅O₇S MW: 627.54 g/mol |
MDL Number:
MFCD00059539 CAS Number: 2429-82-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...