Dabsyl-L-valine ≥98.0% (by HPLC)
Supplier: TCI America
Synonyms:
4-Dimethylaminoazobenzene-4'-sulfonyl-L-valine
D1460-100MG
TCD1460-100MGEA
143
USD
TCD1460-100MG
Dabsyl-L-valine ≥98.0% (by HPLC)
Dabsyl-L-valine
CAS Number: 89131-11-3
MDL Number: MFCD00059366
Molecular Formula: C19H24N4O4S
Molecular Weight: 404.49
Purity/Analysis Method:
Form: Crystal
Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: 4-Dimethylaminoazobenzene-4'-sulfonyl-L-valine, Dbs-Val-OH
Formula:
C₁₉H₂₄N₄O₄S MW: 404.49 g/mol |
MDL Number:
MFCD00059366 CAS Number: 89131-11-3 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...