Dabsyl-L-tryptophan ≥98.0% (by HPLC)

Supplier: TCI America

Synonyms: 4-Dimethylaminoazobenzene-4'-sulfonyl-L-tryptophanDbs-Trp-OH

D1459-100MG
TCD1459-100MGEA 146 USD
TCD1459-100MG
Dabsyl-L-tryptophan ≥98.0% (by HPLC)
Dabsyl-L-tryptophan

CAS Number: 97685-00-2
MDL Number: MFCD00059700
Molecular Formula: C25H25N5O4S
Molecular Weight: 491.57
Purity/Analysis Method: 98.0% (HPLC)
Form: Crystal
Melting point (°C): 206


Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: 4-Dimethylaminoazobenzene-4'-sulfonyl-L-tryptophan, Dbs-Trp-OH

Formula: C₂₅H₂₅N₅O₄S
MW: 491.57 g/mol
Melting Pt: 206 °C
MDL Number: MFCD00059700
CAS Number: 97685-00-2
https://us.vwr.com/stibo/web/std.lang.all/82/91/28978291.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR