Dabsyl-L-proline ≥98.0% (by HPLC)

Supplier: TCI America

Synonyms: 4-Dimethylaminoazobenzene-4'-sulfonyl-L-prolineDbs-Pro-OH

D1458-100MG
TCD1458-100MGEA 146 USD
TCD1458-100MG
Dabsyl-L-proline ≥98.0% (by HPLC)
Dabsyl-L-proline

CAS Number: 89131-09-9
MDL Number: MFCD00059729
Molecular Formula: C19H22N4O4S
Molecular Weight: 402.47
Purity/Analysis Method: 98.0% (HPLC)
Form: Crystal
Melting point (°C): 172


Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: 4-Dimethylaminoazobenzene-4'-sulfonyl-L-proline, Dbs-Pro-OH

Formula: C₁₉H₂₂N₄O₄S
MW: 402.47 g/mol
Melting Pt: 172 °C
MDL Number: MFCD00059729
CAS Number: 89131-09-9
https://us.vwr.com/stibo/web/std.lang.all/82/90/28978290.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR