Dabsyl-L-leucine ≥98.0% (by HPLC)

Supplier: TCI America

Synonyms: 4-Dimethylaminoazobenzene-4'-sulfonyl-L-leucineDbs-Leu-OH

D1456-100MG
TCD1456-100MGEA 146 USD
TCD1456-100MG
Dabsyl-L-leucine ≥98.0% (by HPLC)
Dabsyl-L-leucine

CAS Number: 89131-12-4
MDL Number: MFCD00059354
Molecular Formula: C20H26N4O4S
Molecular Weight: 418.51
Purity/Analysis Method: 98.0% (HPLC)
Form: Crystal
Melting point (°C): 192


Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: 4-Dimethylaminoazobenzene-4'-sulfonyl-L-leucine, Dbs-Leu-OH

Formula: C₂₀H₂₆N₄O₄S
MW: 418.51 g/mol
Melting Pt: 192 °C
MDL Number: MFCD00059354
CAS Number: 89131-12-4
https://us.vwr.com/stibo/web/std.lang.all/82/86/28978286.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR