Dabsyl-L-alanine ≥95.0% (by HPLC)
Synonyms:
4-Dimethylaminoazobenzene-4'-sulfonyl-L-alanine
D1454-100MG
TCD1454-100MGEA
147.29
USD
TCD1454-100MG
Dabsyl-L-alanine ≥95.0% (by HPLC)
Dabsyl-L-alanine
CAS Number: 89131-10-2
MDL Number: MFCD00059648
Molecular Formula: C17H20N4O4S
Molecular Weight: 376.43
Purity/Analysis Method:
Form: Crystal
Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: 4-Dimethylaminoazobenzene-4'-sulfonyl-L-alanine, Dbs-Ala-OH
Formula:
C₁₇H₂₀N₄O₄S MW: 376.43 g/mol |
MDL Number:
MFCD00059648 CAS Number: 89131-10-2 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...