2,5-Dimethyl-3-hexanone ≥96.0%

Supplier: TCI America

D1453-5ML
TCD1453-005MLEA 201 USD
TCD1453-005ML
2,5-Dimethyl-3-hexanone ≥96.0%
2,5-Dimethyl-3-hexanone

CAS Number: 1888-57-9
MDL Number: MFCD00048317
Molecular Formula: C8H16O
Molecular Weight: 128.22
Purity/Analysis Method: 96.0% (GC)
Form: Clear Liquid
Boiling point (°C): 148
Flash Point (°C): 38
Specific Gravity (20/20): 0.81


Other Product Names / Synonyms: Isobutyl Isopropyl Ketone

Formula: C₈H₁₆O
Boiling Pt: 148 °C
Density: 0.81 (20 °C)
Flash Pt: 38 °C
MDL Number: MFCD00048317
CAS Number: 1888-57-9

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR