Dansyl acid ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
Synonyms:
5-Dimethylaminonaphthalene-1-sulfonic Acid
, 5-Dimethylaminonaphthalene-1-sulphonic Acid
D1426-25G
TCD1426-025GEA
182.25
USD
TCD1426-025G
Dansyl acid ≥98.0% (by HPLC, titration analysis)
Dansyl acid
CAS Number: 4272-77-9
MDL Number: MFCD00003995
Molecular Formula: C12H13NO3S
Molecular Weight: 251.30
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 316
Other Product Names / Synonyms: 5-Dimethylaminonaphthalene-1-sulfonic Acid
Formula:
C₁₂H₁₃NO₃S MW: 251.30 g/mol Melting Pt: 316 °C |
MDL Number:
MFCD00003995 CAS Number: 4272-77-9 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...