Dimedone dioxime

Supplier: TCI America

Synonyms: 5,5-Dimethyl-1,3-cyclohexanedione DioximeDimethyldihydroresorcinol DioximeMethone Dioxime

D1400-1G
TCD1400-001GEA 103.48 USD
TCD1400-001G
Dimedone dioxime
Dimedone dioxime

[for Determination of Co]
CAS Number: 37110-24-0
MDL Number: MFCD00059877
Molecular Formula: C8H14N2O2
Molecular Weight: 170.21
Form: Crystal
Color: White
Melting point (°C): 173


Other Product Names / Synonyms: 5,5-Dimethyl-1,3-cyclohexanedione Dioxime, Dimethyldihydroresorcinol Dioxime, Methone Dioxime

Formula: C₈H₁₄N₂O₂
MW: 170.21 g/mol
Melting Pt: 173 °C
MDL Number: MFCD00059877
CAS Number: 37110-24-0
https://us.vwr.com/stibo/web/std.lang.all/82/47/28978247.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR