(-)-Diacetyl-L-tartaric acid ≥98.0% (by titrimetric analysis)

Supplier: TCI America

D1386-25G
TCD1386-025GEA 181.61 USD
TCD1386-025G
(-)-Diacetyl-L-tartaric acid ≥98.0% (by titrimetric analysis)
(-)-Diacetyl-L-tartaric acid

CAS Number: 51591-38-9
MDL Number: MFCD00070579
Molecular Formula: C8H10O8
Molecular Weight: 234.16
Purity/Analysis Method: 98.0% (T)
Form: Crystal
Melting point (°C): 120
Specific rotation [a]20/D: -25 deg (C=10, Acetone)

Formula: C₈H₁₀O₈
Melting Pt: 120 °C
MDL Number: MFCD00070579
CAS Number: 51591-38-9
https://us.vwr.com/stibo/web/std.lang.all/82/35/28978235.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR