Furfuryl disulfide ≥95.0%

Supplier: TCI America

Synonyms: 1,2-Bis(furan-2-ylmethyl)disulfane1,2-Bis(furan-2-ylmethyl)disulphane

D1384-25G
TCD1384-025GEA 68.49 USD
TCD1384-025G
Furfuryl disulfide ≥95.0%
Furfuryl disulphide

CAS Number: 4437-20-1
MDL Number: MFCD00010082
Molecular Formula: C10H10O2S2
Molecular Weight: 226.31
Purity/Analysis Method: 95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 121
Flash Point (°C): 113
Specific Gravity (20/20): 1.24
Storage Temperature: 0-10°C


Other Product Names / Synonyms: Furfuryl Disulfide

Formula: C₁₀H₁₀O₂S₂
MW: 226.3152 g/mol
Boiling Pt: 121 °C
Density: 1.24 (20 °C)
Flash Pt: 113 °C
MDL Number: MFCD00010082
CAS Number: 4437-20-1
https://us.vwr.com/stibo/web/std.lang.all/82/34/28978234.jpg

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR