5-Decanol ≥96.0%

Supplier: TCI America

D1381-5ML
TCD1381-005MLEA 182.64 USD
TCD1381-005ML
5-Decanol ≥96.0%
5-Decanol

CAS Number: 5205-34-5
MDL Number: MFCD00039626
Molecular Formula: C10H22O
Molecular Weight: 158.29
Purity/Analysis Method: 96.0% (GC)
Form: Clear Liquid
Boiling point (°C): 80
Melting point (°C): 9
Specific Gravity (20/20): 0.83


Other Product Names / Synonyms: Amylbutylcarbinol, Butylpentylcarbinol, 5-Decyl Alcohol

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR