Diethylamine acetate ≥97.0% (by titrimetric analysis)

Supplier: TCI America

D1352-25G
TCD1352-025GEA 108.57 USD
TCD1352-025G
Diethylamine acetate ≥97.0% (by titrimetric analysis)
Diethylamine acetate

CAS Number: 20726-63-0
MDL Number: MFCD00060243
Molecular Formula: C4H11N
Molecular Weight: 133.19
Purity/Analysis Method: 97.0% (T)
Form: Crystal
Melting point (°C): 47

Formula: C₄H₁₁N
Melting Pt: 47 °C
MDL Number: MFCD00060243
CAS Number: 20726-63-0
https://us.vwr.com/stibo/web/std.lang.all/82/15/28978215.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR