1,1-Dimethyl-4-phenylpiperazinium iodide ≥98.0% (by HPLC)
D1343-1G
TCD1343-001GEA
59.94
USD
TCD1343-001G
1,1-Dimethyl-4-phenylpiperazinium iodide ≥98.0% (by HPLC)
1,1-Dimethyl-4-phenylpiperazinium iodide
CAS Number: 54-77-3
MDL Number: MFCD00011976
Molecular Formula: C12H19IN2
Molecular Weight: 318.20
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 232
Other Product Names / Synonyms: 1-Methyl-4-phenylpiperazinium 1-Methiodide
Formula:
C₁₂H₁₉IN₂ Melting Pt: 232 °C |
MDL Number:
MFCD00011976 CAS Number: 54-77-3 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...