Di-p-tolylmethane ≥99.0%

Supplier: TCI America

Synonyms: 4,4'-Dimethyldiphenylmethane

D1312-25ML
TCD1312-025MLEA 272 USD
TCD1312-025ML
Di-p-tolylmethane ≥99.0%
Di-p-tolylmethane

CAS Number: 4957-14-6
MDL Number: MFCD00026018
Molecular Formula: C15H16
Molecular Weight: 196.29
Purity/Analysis Method: 99.0% (GC)
Form: Crystal
Boiling point (°C): 150
Freezing point (°C): 28


Other Product Names / Synonyms: 4,4'-Dimethyldiphenylmethane

Formula: C₁₅H₁₆
MW: 196.29 g/mol
Boiling Pt: 150 °C
MDL Number: MFCD00026018
CAS Number: 4957-14-6
https://us.vwr.com/stibo/web/std.lang.all/81/84/28978184.jpg

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR