N,N-Diethyl-N'-methylethylenediamine ≥98.0% (by GC, titration analysis)

Supplier: TCI America

D1297-5ML D1297-25ML
TCD1297-005MLEA 47.71 USD
TCD1297-005ML TCD1297-025ML
N,N-Diethyl-N'-methylethylenediamine ≥98.0% (by GC, titration analysis)
N,N-Diethyl-N'-methylethylenediamine

CAS Number: 104-79-0
MDL Number: MFCD00009053
Molecular Formula: C7H18N2
Molecular Weight: 130.24
Purity/Analysis Method: 98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 156
Flash Point (°C): 41
Specific Gravity (20/20): 0.81

Formula: C₇H₁₈N₂
Boiling Pt: 156 °C
Density: 0.81 (20 °C)
Flash Pt: 41 °C
MDL Number: MFCD00009053
CAS Number: 104-79-0
https://us.vwr.com/stibo/web/std.lang.all/81/76/28978176.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR