3,4-Dimethylcyclohexanone ≥98.0%

Supplier: TCI America

D1281-10ML
TCD1281-010MLEA 402.96 USD
TCD1281-010ML
3,4-Dimethylcyclohexanone ≥98.0%
3,4-Dimethylcyclohexanone

CAS Number: 5465-09-8
MDL Number: MFCD00045552
Molecular Formula: C8H14O
Molecular Weight: 126.20
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Flash Point (°C): 51
Specific Gravity (20/20): 0.92

Formula: C₈H₁₄O
Density: 0,92 (20 °C)
Flash Pt: 51 °C
MDL Number: MFCD00045552
CAS Number: 5465-09-8

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR