Acridinium chloride ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: Acridine hydrochloride

A0131-5G
TCA0131-5GEA 214.06 USD
TCA0131-5G
Acridinium chloride ≥98.0% (by HPLC, titration analysis)
Acridinium chloride

CAS Number: 17784-47-3
MDL Number: MFCD00035149
Molecular Formula: C13H9N
Molecular Weight: 215.68
Purity/Analysis Method: 98.0% (HPLC,T)
Form: Crystal

https://us.vwr.com/stibo/web/std.lang.all/75/78/28927578.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR